Product Details

SNP ID
rs143742503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32950576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCTATTTGCTGGATCATCCACT[C/T]GCAGAACTCTTTGTCAAATAAAATG
Phenotype
MIM: 142855 MIM: 142856
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HLA-DMA PubMed Links

Gene Details

Gene
HLA-DMA
Gene Name
major histocompatibility complex, class II, DM alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006120.3 402 Missense Mutation AAG,GAG K106E NP_006111.2
Gene
HLA-DMB
Gene Name
major histocompatibility complex, class II, DM beta
There are no transcripts associated with this gene.

View Full Product Details