Product Details

SNP ID
rs143962430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167847141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTTTCATGCTGACACATCCAGT[A/C]GTTTATCTTCTGATGCAGCAGCAGT
Phenotype
MIM: 159559
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AFDN PubMed Links

Gene Details

Gene
AFDN
Gene Name
afadin, adherens junction formation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040000.2 Intron NP_001035089.1
NM_001207008.1 Intron NP_001193937.1
NM_001291964.1 Intron NP_001278893.1
XM_005266996.3 Intron XP_005267053.1
XM_005266997.2 Intron XP_005267054.1
XM_006715491.2 Intron XP_006715554.1
XM_006715494.3 Intron XP_006715557.1
XM_011535842.2 Intron XP_011534144.1
XM_017010878.1 Intron XP_016866367.1
XM_017010879.1 Intron XP_016866368.1
XM_017010880.1 Intron XP_016866369.1
XM_017010881.1 Intron XP_016866370.1
XM_017010882.1 Intron XP_016866371.1
XM_017010883.1 Intron XP_016866372.1
XM_017010884.1 Intron XP_016866373.1
XM_017010885.1 Intron XP_016866374.1
XM_017010886.1 Intron XP_016866375.1
XM_017010887.1 Intron XP_016866376.1

View Full Product Details