Product Details

SNP ID
rs143964760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30578130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCGGAAGAAGGATGTGGATGAT[G/T]ATGGAGAAGAGAAAGAGCTCATGGA
Phenotype
MIM: 603429 MIM: 611619
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCF1 PubMed Links

Gene Details

Gene
ABCF1
Gene Name
ATP binding cassette subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025091.1 366 Missense Mutation GAT,TAT D91Y NP_001020262.1
NM_001090.2 366 Missense Mutation GAT,TAT D91Y NP_001081.1
Gene
MIR877
Gene Name
microRNA 877
There are no transcripts associated with this gene.

View Full Product Details