Product Details

SNP ID
rs144004432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:139148081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATCTTTTTATATTTATATTGATC[A/G]TTTGTATTTTTTTCTTGTTTGTGAT
Phenotype
MIM: 607977
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HECA PubMed Links

Gene Details

Gene
HECA
Gene Name
hdc homolog, cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016217.2 Intron NP_057301.1

View Full Product Details