Product Details

SNP ID
rs144136339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30949051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGGAAAAGTCCATGAGTACTT[C/T]GGATAAGACAAGTACCAGCTCACAT
Phenotype
MIM: 613928
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPCR1 PubMed Links

Gene Details

Gene
DPCR1
Gene Name
diffuse panbronchiolitis critical region 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080870.3 587 Missense Mutation TCG,TTG S196L NP_543146.2

View Full Product Details