Product Details

SNP ID
rs144230168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:79545618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCGTTTACAATTCCCTCTCACTA[C/T]ATGTATATAAGTAACTATTAGGCTA
Phenotype
MIM: 611408 MIM: 615678
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCA5 PubMed Links

Gene Details

Gene
LCA5
Gene Name
LCA5, lebercilin
There are no transcripts associated with this gene.

Gene
SH3BGRL2
Gene Name
SH3 domain binding glutamate rich protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031469.3 Intron NP_113657.1
XM_011536165.2 Intron XP_011534467.1
XM_011536282.2 Intron XP_011534584.1
XM_017011344.1 Intron XP_016866833.1
XM_017011345.1 Intron XP_016866834.1

View Full Product Details