Product Details

SNP ID
rs144338608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:118814839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCCCAACTATGACTTTTTTCT[C/T]ATCTCTTCATCCCAGTCACAATCAA
Phenotype
MIM: 610098
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MCM9 PubMed Links

Gene Details

Gene
MCM9
Gene Name
minichromosome maintenance 9 homologous recombination repair factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017696.2 3432 Missense Mutation ATA,ATG I1139M NP_060166.2
NM_153255.4 3432 Intron NP_694987.1

View Full Product Details