Product Details

SNP ID
rs144491725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:34793425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGATGTCGGTCAACATGGGACAC[C/T]CAGGCTTGCACTGATGTTTCTTTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UHRF1BP1 PubMed Links

Gene Details

Gene
UHRF1BP1
Gene Name
UHRF1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017754.3 Intron NP_060224.3

View Full Product Details