Product Details

SNP ID
rs113030936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30730049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGATGCTCACGCCTGTGAGTCT[C/T]TCCACACTCTCTGGCAGGCGAGTTA
Phenotype
MIM: 606998 MIM: 191130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FLOT1 PubMed Links

Gene Details

Gene
FLOT1
Gene Name
flotillin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318875.1 1566 Silent Mutation GAA,GAG E361E NP_001305804.1
NM_005803.3 1566 Silent Mutation GAA,GAG E409E NP_005794.1
XM_005248780.3 1566 Silent Mutation GAA,GAG E409E XP_005248837.1
XM_006714947.3 1566 Silent Mutation GAA,GAG E356E XP_006715010.1
XM_017010157.1 1566 Silent Mutation GAA,GAG E405E XP_016865646.1
XM_017010158.1 1566 Silent Mutation GAA,GAG E372E XP_016865647.1
Gene
TUBB
Gene Name
tubulin beta class I
There are no transcripts associated with this gene.

View Full Product Details