Product Details

SNP ID
rs144753674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:83857271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGGCCAAAATCAAAATGTTATG[A/T]TTACTTATATCAAGAAGCAGAAGCT
Phenotype
MIM: 608343 MIM: 609891
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYB5R4 PubMed Links

Gene Details

Gene
CYB5R4
Gene Name
cytochrome b5 reductase 4
There are no transcripts associated with this gene.

Gene
RIPPLY2
Gene Name
ripply transcriptional repressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009994.2 420 Missense Mutation GAT,GTT D90V NP_001009994.1

View Full Product Details