Product Details

SNP ID
rs144796171
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.6:30163747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGCCTGTACCCTCTGTGCGGG[A/G]CCGCTGGAGGATGCGGTGACCATTC
Phenotype
MIM: 605701
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TRIM10 PubMed Links

Gene Details

Gene
TRIM10
Gene Name
tripartite motif containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006778.3 542 Intron NP_006769.2
NM_052828.2 542 Intron NP_439893.2
XM_011514221.1 542 Intron XP_011512523.1
XM_011514222.2 542 Intron XP_011512524.1
XM_011514223.2 542 Intron XP_011512525.1
XM_011514225.1 542 Intron XP_011512527.1
Gene
TRIM15
Gene Name
tripartite motif containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033229.2 542 Silent Mutation GGA,GGG G21G NP_150232.2
XM_011514987.1 542 Intron XP_011513289.1
XM_011514988.2 542 UTR 5 XP_011513290.1

View Full Product Details