Product Details

SNP ID
rs144832798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169751382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAGGCCTCCTTTCTTTCCTAGGC[A/G]TCACCGGAGCCTGCTGAGGGGTCTG
Phenotype
MIM: 615532 MIM: 186977
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERMARD PubMed Links

Gene Details

Gene
ERMARD
Gene Name
ER membrane associated RNA degradation
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278531.1 200 Intron NP_001265460.1
NM_001278532.1 200 Intron NP_001265461.1
NM_001278533.1 200 Intron NP_001265462.1
NM_018341.2 200 Intron NP_060811.1
XM_011535938.2 200 Intron XP_011534240.1
XM_011535939.2 200 Intron XP_011534241.1
XM_011535940.2 200 Intron XP_011534242.1
XM_017011027.1 200 Intron XP_016866516.1
XM_017011028.1 200 Intron XP_016866517.1
XM_017011029.1 200 Intron XP_016866518.1
XM_017011030.1 200 Intron XP_016866519.1
XM_017011031.1 200 Intron XP_016866520.1
XM_017011032.1 200 Intron XP_016866521.1
XM_017011033.1 200 Intron XP_016866522.1
Gene
TCTE3
Gene Name
t-complex-associated-testis-expressed 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006715554.3 200 Missense Mutation ACG,ATG T26M XP_006715617.1
XM_011536093.2 200 Missense Mutation ACG,ATG T26M XP_011534395.1

View Full Product Details