Product Details

SNP ID
rs145203673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:25725328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTTTTTGACATATTTTACTTA[C/T]ATTTTAAGAAATAATTCTATTATTC
Phenotype
MIM: 613499 MIM: 609904 MIM: 182308
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIST1H2AA PubMed Links

Gene Details

Gene
HIST1H2AA
Gene Name
histone cluster 1, H2aa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_170745.3 Intron NP_734466.1
Gene
HIST1H2APS1
Gene Name
histone cluster 1, H2a, pseudogene 1
There are no transcripts associated with this gene.

Gene
HIST1H2BA
Gene Name
histone cluster 1, H2ba
There are no transcripts associated with this gene.

Gene
SLC17A1
Gene Name
solute carrier family 17 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005074.3 Intron NP_005065.2
XM_011514818.2 Intron XP_011513120.2
XM_011514819.2 Intron XP_011513121.2
XM_011514820.2 Intron XP_011513122.2
XM_011514821.2 Intron XP_011513123.1
XM_017011199.1 Intron XP_016866688.1
XM_017011200.1 Intron XP_016866689.1
XM_017011201.1 Intron XP_016866690.1
XM_017011202.1 Intron XP_016866691.1

View Full Product Details