Product Details

SNP ID
rs6926628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32949721 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAAGAAAAGGCCTGGAAGCTGAGT[C/G]CATCGACAGCTGAGACAAAAGTAGG
Phenotype
MIM: 142855 MIM: 142856
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HLA-DMA PubMed Links

Gene Details

Gene
HLA-DMA
Gene Name
major histocompatibility complex, class II, DM alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006120.3 628 Missense Mutation GCA,GGA A181G NP_006111.2
Gene
HLA-DMB
Gene Name
major histocompatibility complex, class II, DM beta
There are no transcripts associated with this gene.

View Full Product Details