Product Details

SNP ID
rs145232088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:158113570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCAAACTCCAGAAAGTCATCTTG[C/T]AGTGTTTTAAGTGCAGGAGAATCTG
Phenotype
MIM: 614725
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERAC1 PubMed Links

Gene Details

Gene
SERAC1
Gene Name
serine active site containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032861.3 1774 Silent Mutation CTA,CTG L569L NP_116250.3
XM_006715586.2 1774 Silent Mutation CTA,CTG L499L XP_006715649.1
XM_011536196.2 1774 Silent Mutation CTA,CTG L562L XP_011534498.1
XM_011536198.2 1774 Silent Mutation CTA,CTG L499L XP_011534500.1

View Full Product Details