Product Details

SNP ID
rs145391320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:132617450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTTTTTTGTCTTTCTTCACTTG[A/G]TTATTTTGATTTTCTCGCAAGTTAT
Phenotype
MIM: 604849
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAAR2 PubMed Links

Gene Details

Gene
TAAR2
Gene Name
trace amine associated receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033080.1 644 Silent Mutation AAC,AAT N252N NP_001028252.1
NM_014626.3 644 Silent Mutation AAC,AAT N207N NP_055441.2
Gene
TAAR3
Gene Name
trace amine associated receptor 3 (gene/pseudogene)
There are no transcripts associated with this gene.

View Full Product Details