Product Details

SNP ID
rs145569691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:152971338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTTTGCAGGTTGCCCGTTGTAAA[C/T]AGCAATCATATTTTGCAGGTGAATT
Phenotype
MIM: 606013
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO5 PubMed Links

Gene Details

Gene
FBXO5
Gene Name
F-box protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142522.1 1278 Missense Mutation TAT,TGT Y344C NP_001135994.1
NM_012177.3 1278 Missense Mutation TAT,TGT Y390C NP_036309.1

View Full Product Details