Product Details

SNP ID
rs145902743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:139166649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGGCTCCGAGAAGAACACAGGG[A/G]GGCCTCCTGGTGAGGCGGCGGAGGA
Phenotype
MIM: 607977 MIM: 611438
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HECA PubMed Links

Gene Details

Gene
HECA
Gene Name
hdc homolog, cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016217.2 922 Missense Mutation AGG,GGG R213G NP_057301.1
Gene
TXLNB
Gene Name
taxilin beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153235.3 922 Intron NP_694967.3
XM_005266836.1 922 Intron XP_005266893.1
XM_011535505.2 922 Intron XP_011533807.1
XM_011535506.2 922 Intron XP_011533808.1
XM_017010322.1 922 Intron XP_016865811.1

View Full Product Details