Product Details

SNP ID
rs145917056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42228054 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGACATCTGAATGCAATGGAACA[C/T]GAAGGTCAGCTTCAGTCCCTACTGG
Phenotype
MIM: 610322
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRERF1 PubMed Links

Gene Details

Gene
TRERF1
Gene Name
transcriptional regulating factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297573.1 4941 UTR 3 NP_001284502.1
NM_033502.3 4941 UTR 3 NP_277037.1
XM_011514741.2 4941 UTR 3 XP_011513043.1
XM_011514742.2 4941 UTR 3 XP_011513044.1
XM_011514743.2 4941 UTR 3 XP_011513045.1
XM_011514744.1 4941 UTR 3 XP_011513046.1
XM_011514745.1 4941 UTR 3 XP_011513047.1
XM_017011034.1 4941 UTR 3 XP_016866523.1
XM_017011035.1 4941 UTR 3 XP_016866524.1
XM_017011036.1 4941 UTR 3 XP_016866525.1
XM_017011037.1 4941 UTR 3 XP_016866526.1
XM_017011038.1 4941 UTR 3 XP_016866527.1
XM_017011039.1 4941 UTR 3 XP_016866528.1
XM_017011040.1 4941 UTR 3 XP_016866529.1
XM_017011041.1 4941 UTR 3 XP_016866530.1
XM_017011042.1 4941 UTR 3 XP_016866531.1
XM_017011043.1 4941 UTR 3 XP_016866532.1
XM_017011044.1 4941 UTR 3 XP_016866533.1
XM_017011045.1 4941 UTR 3 XP_016866534.1
XM_017011046.1 4941 UTR 3 XP_016866535.1
XM_017011047.1 4941 UTR 3 XP_016866536.1
XM_017011048.1 4941 UTR 3 XP_016866537.1
XM_017011049.1 4941 UTR 3 XP_016866538.1
XM_017011050.1 4941 UTR 3 XP_016866539.1
XM_017011051.1 4941 UTR 3 XP_016866540.1
XM_017011052.1 4941 UTR 3 XP_016866541.1
XM_017011053.1 4941 UTR 3 XP_016866542.1
XM_017011054.1 4941 UTR 3 XP_016866543.1
XM_017011055.1 4941 UTR 3 XP_016866544.1

View Full Product Details