Product Details

SNP ID
rs145995335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33398130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAGGAAGCAGACTCAAGAGGAG[A/G]CCTGACCAGATGGAAGATGGCCTGG
Phenotype
MIM: 603763
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIFC1 PubMed Links

Gene Details

Gene
KIFC1
Gene Name
kinesin family member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002263.3 270 Silent Mutation AGA,AGG R38R NP_002254.2
XM_011514585.1 270 Silent Mutation AGA,AGG R38R XP_011512887.1
XM_011514587.2 270 Silent Mutation AGA,AGG R38R XP_011512889.1
XM_017010836.1 270 Silent Mutation AGA,AGG R38R XP_016866325.1
XM_017010837.1 270 UTR 5 XP_016866326.1

View Full Product Details