Product Details

SNP ID
rs145998938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:114055652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCATCAAATCACTAGACAGGAT[A/G]AACAACTCATGGAGAAAATCAACAC
Phenotype
MIM: 609407
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HS3ST5 PubMed Links

Gene Details

Gene
HS3ST5
Gene Name
heparan sulfate-glucosamine 3-sulfotransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153612.3 4336 UTR 3 NP_705840.2
XM_006715379.2 4336 UTR 3 XP_006715442.1
XM_011535588.2 4336 UTR 3 XP_011533890.1
XM_017010470.1 4336 UTR 3 XP_016865959.1
XM_017010471.1 4336 UTR 3 XP_016865960.1
XM_017010472.1 4336 UTR 3 XP_016865961.1
XM_017010473.1 4336 UTR 3 XP_016865962.1
XM_017010474.1 4336 UTR 3 XP_016865963.1
Gene
LOC101927768
Gene Name
uncharacterized LOC101927768
There are no transcripts associated with this gene.

View Full Product Details