Product Details

SNP ID
rs146067319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26225245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTAAGGCAGCTCGCAAGAGCGCT[C/T]CGGCCACGGGCGGCGTGAAGAAGCC
Phenotype
MIM: 142210 MIM: 602786 MIM: 602798 MIM: 602813
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIST1H1D PubMed Links

Gene Details

Gene
HIST1H1D
Gene Name
histone cluster 1, H1d
There are no transcripts associated with this gene.

Gene
HIST1H2AE
Gene Name
histone cluster 1, H2ae
There are no transcripts associated with this gene.

Gene
HIST1H2BG
Gene Name
histone cluster 1, H2bg
There are no transcripts associated with this gene.

Gene
HIST1H3E
Gene Name
histone cluster 1, H3e
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003532.2 91 Missense Mutation CCG,TCG P31S NP_003523.1

View Full Product Details