Product Details

SNP ID
rs146219124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:17760810 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTGCTTGCCCAAGGTGACCAGG[C/T]GAACAAAGACGCCAAGCTGTGGCCT
Phenotype
MIM: 605433
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF13A PubMed Links

Gene Details

Gene
KIF13A
Gene Name
kinesin family member 13A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105566.2 5596 Intron NP_001099036.1
NM_001105567.2 5596 Intron NP_001099037.1
NM_001105568.2 5596 UTR 3 NP_001099038.1
NM_001243423.1 5596 Intron NP_001230352.1
NM_022113.5 5596 Intron NP_071396.4
XM_011514806.2 5596 Intron XP_011513108.1
XM_017011188.1 5596 UTR 3 XP_016866677.1
XM_017011189.1 5596 Intron XP_016866678.1
XM_017011190.1 5596 Intron XP_016866679.1
XM_017011191.1 5596 Intron XP_016866680.1
XM_017011192.1 5596 UTR 3 XP_016866681.1
XM_017011193.1 5596 Intron XP_016866682.1

View Full Product Details