Product Details

SNP ID
rs146329157
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110612205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTCCTTGAGAGCAAGAACAGAC[C/T]TTCTATTTTCCTTGCAATTTCCTAC
Phenotype
MIM: 614720
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK19 PubMed Links

Gene Details

Gene
CDK19
Gene Name
cyclin dependent kinase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300960.1 3602 UTR 3 NP_001287889.1
NM_001300963.1 3602 UTR 3 NP_001287892.1
NM_001300964.1 3602 UTR 3 NP_001287893.1
NM_015076.4 3602 UTR 3 NP_055891.1
XM_005266871.3 3602 UTR 3 XP_005266928.1
XM_011535630.2 3602 UTR 3 XP_011533932.1
XM_011535631.2 3602 UTR 3 XP_011533933.1
XM_011535632.1 3602 Intron XP_011533934.1
XM_017010587.1 3602 UTR 3 XP_016866076.1
XM_017010588.1 3602 UTR 3 XP_016866077.1

View Full Product Details