Product Details

SNP ID
rs146503956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:26642789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGGACTTATACTGGGGCTGGAT[C/T]CCAGATCTACTCCACCTATCACCTG
Phenotype
MIM: 610847
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF322 PubMed Links

Gene Details

Gene
ZNF322
Gene Name
zinc finger protein 322
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242797.1 Intron NP_001229726.1
NM_001242798.1 Intron NP_001229727.1
NM_001242799.1 Intron NP_001229728.1
NM_024639.4 Intron NP_078915.2

View Full Product Details