Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000125.3 | 1180 | Intron | NP_000116.2 | ||
NM_001122740.1 | 1180 | Intron | NP_001116212.1 | ||
NM_001122741.1 | 1180 | Intron | NP_001116213.1 | ||
NM_001122742.1 | 1180 | Intron | NP_001116214.1 | ||
NM_001291230.1 | 1180 | Intron | NP_001278159.1 | ||
NM_001291241.1 | 1180 | Intron | NP_001278170.1 | ||
NM_001328100.1 | 1180 | Intron | NP_001315029.1 | ||
XM_006715374.3 | 1180 | Intron | XP_006715437.1 | ||
XM_006715375.3 | 1180 | Intron | XP_006715438.1 | ||
XM_011535543.2 | 1180 | Intron | XP_011533845.1 | ||
XM_011535544.2 | 1180 | Intron | XP_011533846.1 | ||
XM_011535545.2 | 1180 | Intron | XP_011533847.1 | ||
XM_011535547.2 | 1180 | Intron | XP_011533849.1 | ||
XM_011535549.2 | 1180 | Intron | XP_011533851.1 | ||
XM_017010376.1 | 1180 | UTR 5 | XP_016865865.1 | ||
XM_017010377.1 | 1180 | UTR 5 | XP_016865866.1 | ||
XM_017010378.1 | 1180 | Intron | XP_016865867.1 | ||
XM_017010379.1 | 1180 | Intron | XP_016865868.1 | ||
XM_017010380.1 | 1180 | UTR 5 | XP_016865869.1 | ||
XM_017010381.1 | 1180 | Intron | XP_016865870.1 | ||
XM_017010382.1 | 1180 | Intron | XP_016865871.1 | ||
XM_017010383.1 | 1180 | Intron | XP_016865872.1 |