Product Details

SNP ID
rs146671770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43178411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCTCGGGCCTGGGTGATGGCA[A/G]CCTCACCGTGCTGAATGCCTTCTCC
Phenotype
MIM: 607489 MIM: 600589
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUL9 PubMed Links

Gene Details

Gene
CUL9
Gene Name
cullin 9
There are no transcripts associated with this gene.

Gene
SRF
Gene Name
serum response factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292001.1 1642 Missense Mutation AAC,AGC N223S NP_001278930.1
NM_003131.3 1642 Missense Mutation AAC,AGC N427S NP_003122.1

View Full Product Details