Product Details

SNP ID
rs147031652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41739922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCAACCCTCAGAACACCAGCC[A/G]GAGGCCTGGCCGCCGATGAGGAACC
Phenotype
MIM: 169740 MIM: 600744
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PGC PubMed Links

Gene Details

Gene
PGC
Gene Name
progastricsin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166424.1 859 Intron NP_001159896.1
NM_002630.3 859 Silent Mutation TCC,TCT S264S NP_002621.1
Gene
TFEB
Gene Name
transcription factor EB
There are no transcripts associated with this gene.

View Full Product Details