Product Details

SNP ID
rs147098824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89364123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTCATCTTTTTGACCTTGGTTG[C/T]CTCATCTGAAAAATAGAAATGGTAA
Phenotype
MIM: 608268
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RRAGD PubMed Links

Gene Details

Gene
RRAGD
Gene Name
Ras related GTP binding D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021244.4 5464 Intron NP_067067.1
XM_005248755.4 5464 UTR 3 XP_005248812.1
XM_011536022.2 5464 Intron XP_011534324.1

View Full Product Details