Product Details

SNP ID
rs147188374
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46702084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGTGTTTATAGTGATCCTGTTA[C/T]ATATACAGATCTGGGATCTTTCGTC
Phenotype
MIM: 601690 MIM: 611200
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G7 PubMed Links

Gene Details

Gene
PLA2G7
Gene Name
phospholipase A2 group VII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168357.1 6652 Intron NP_001161829.1
NM_005084.3 6652 Intron NP_005075.3
XM_005249408.4 6652 Intron XP_005249465.1
Gene
TDRD6
Gene Name
tudor domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010870.2 6652 UTR 3 NP_001010870.1
NM_001168359.1 6652 UTR 3 NP_001161831.1

View Full Product Details