Product Details

SNP ID
rs147443268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:100389418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGGTTTTCCTTTTATTTTTGCA[C/T]CTATTGGTCTTTTTTCTGGGTGAAT
Phenotype
MIM: 603128
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIM1 PubMed Links

Gene Details

Gene
SIM1
Gene Name
single-minded family bHLH transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005068.2 4027 UTR 3 NP_005059.2
XM_005267100.2 4027 UTR 3 XP_005267157.1
XM_011536072.2 4027 Intron XP_011534374.1
XM_017011197.1 4027 UTR 3 XP_016866686.1

View Full Product Details