Product Details

SNP ID
rs147513433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149578780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCACACTCACTCACAGATTTGGT[A/G]GTAATCTTTGTTCAAAGGTGTCACT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GINM1 PubMed Links

Gene Details

Gene
GINM1
Gene Name
glycoprotein integral membrane 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138785.3 Intron NP_620140.1

View Full Product Details