Product Details

SNP ID
rs147634437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30547416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCAGTCACAGAGCTTCACAGAG[C/G]GGGTAAGAAAGTAGGTCTGAAAGTA
Phenotype
MIM: 143024
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GNL1 PubMed Links

Gene Details

Gene
GNL1
Gene Name
G protein nucleolar 1 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005275.3 2605 Missense Mutation CCC,CGC P405R NP_005266.2
XM_005249015.3 2605 Missense Mutation CCC,CGC P405R XP_005249072.1
Gene
PRR3
Gene Name
proline rich 3
There are no transcripts associated with this gene.

View Full Product Details