Product Details

SNP ID
rs147838633
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:117270566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTGCTCCTTCCCGATGAGCCAG[C/T]GCAGCTTCCCCGCCTCCTTCTGGAA
Phenotype
MIM: 609979
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VGLL2 PubMed Links

Gene Details

Gene
VGLL2
Gene Name
vestigial like family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153453.1 438 Intron NP_703154.1
NM_182645.3 438 Missense Mutation CGC,TGC R139C NP_872586.1
XM_005266883.2 438 Missense Mutation CGC,TGC R139C XP_005266940.1

View Full Product Details