Product Details

SNP ID
rs148039321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43506998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATAAAATCTTTTCCGAGCCAGC[A/G]GGTTGCAGCCAGAGCTGCCAAGTTC
Phenotype
MIM: 612658 MIM: 609775
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC73 PubMed Links

Gene Details

Gene
LRRC73
Gene Name
leucine rich repeat containing 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012974.2 1057 UTR 3 NP_001012992.1
NM_001271882.1 1057 UTR 3 NP_001258811.1
Gene
TJAP1
Gene Name
tight junction associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146016.1 1057 Intron NP_001139488.1
NM_001146017.1 1057 Intron NP_001139489.1
NM_001146018.1 1057 Intron NP_001139490.1
NM_001146019.1 1057 Intron NP_001139491.1
NM_001146020.1 1057 Intron NP_001139492.1
NM_080604.2 1057 Intron NP_542171.2
XM_006715249.1 1057 Intron XP_006715312.1
XM_006715250.3 1057 Intron XP_006715313.1
XM_006715251.2 1057 Intron XP_006715314.1
XM_006715252.2 1057 Intron XP_006715315.1
XM_006715254.3 1057 Intron XP_006715317.1
XM_006715256.1 1057 Intron XP_006715319.1
XM_006715257.2 1057 Intron XP_006715320.1
XM_006715259.3 1057 Intron XP_006715322.1
XM_006715260.1 1057 Intron XP_006715323.1
XM_006715261.1 1057 Intron XP_006715324.1
XM_006715262.2 1057 Intron XP_006715325.1
XM_006715263.2 1057 Intron XP_006715326.1
XM_006715265.1 1057 Intron XP_006715328.1
XM_006715266.1 1057 Intron XP_006715329.1
XM_006715267.2 1057 Intron XP_006715330.1
XM_006715268.2 1057 Intron XP_006715331.1
XM_006715269.2 1057 Intron XP_006715332.1
XM_006715270.1 1057 Intron XP_006715333.1
XM_011514995.1 1057 Intron XP_011513297.1
XM_011514996.1 1057 Intron XP_011513298.1
XM_017011488.1 1057 Intron XP_016866977.1
XM_017011489.1 1057 Intron XP_016866978.1
XM_017011490.1 1057 Intron XP_016866979.1
XM_017011491.1 1057 Intron XP_016866980.1
XM_017011492.1 1057 Intron XP_016866981.1
XM_017011493.1 1057 Intron XP_016866982.1
Gene
YIPF3
Gene Name
Yip1 domain family member 3
There are no transcripts associated with this gene.

View Full Product Details