Product Details

SNP ID
rs148054537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32149251 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATCCCAGTAGTTCTCGTGGTGCT[A/G]CGCGGCGATGATGATGACTACGGTG
Phenotype
MIM: 603298
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100507547 PubMed Links

Gene Details

Gene
LOC100507547
Gene Name
uncharacterized LOC100507547
There are no transcripts associated with this gene.

Gene
PPT2
Gene Name
palmitoyl-protein thioesterase 2
There are no transcripts associated with this gene.

Gene
PPT2-EGFL8
Gene Name
PPT2-EGFL8 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
PRRT1
Gene Name
proline rich transmembrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030651.3 1008 Nonsense Mutation CAG,TAG Q298* NP_085154.3

View Full Product Details