Product Details

SNP ID
rs148218453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36319515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGGTTCCTGTCCAGCTTAGAGGA[A/G]TGAGCCACCTGGCTGCGCAGGGTGG
Phenotype
MIM: 612121
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf222 PubMed Links

Gene Details

Gene
C6orf222
Gene Name
chromosome 6 open reading frame 222
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010903.4 1452 Silent Mutation CAC,CAT H588H NP_001010903.3
XM_011514596.2 1452 Silent Mutation CAC,CAT H588H XP_011512898.1
XM_011514597.2 1452 Silent Mutation CAC,CAT H587H XP_011512899.1
XM_011514598.2 1452 Silent Mutation CAC,CAT H340H XP_011512900.1
Gene
PNPLA1
Gene Name
patatin like phospholipase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details