Product Details

SNP ID
rs148246588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27815056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTTTTCCACCATGCCTGAACCA[A/G]TCAAATCTGCTCCAGTCCCTAAAAA
Phenotype
MIM: 602787 MIM: 602791 MIM: 602800 MIM: 602802 MIM: 602818 MIM: 602826
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIST1H2AI PubMed Links

Gene Details

Gene
HIST1H2AI
Gene Name
histone cluster 1, H2ai
There are no transcripts associated with this gene.

Gene
HIST1H2AJ
Gene Name
histone cluster 1, H2aj
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021066.2 Intron NP_066544.1
Gene
HIST1H2BL
Gene Name
histone cluster 1, H2bl
There are no transcripts associated with this gene.

Gene
HIST1H2BM
Gene Name
histone cluster 1, H2bm
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003521.2 Intron NP_003512.1
Gene
HIST1H3H
Gene Name
histone cluster 1, H3h
There are no transcripts associated with this gene.

Gene
HIST1H4J
Gene Name
histone cluster 1, H4j
There are no transcripts associated with this gene.

View Full Product Details