Product Details
- SNP ID
-
rs148251960
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:42965079 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTCCTCTAACAGAGCATACTTGC[A/G]TGTGATGGCACTTAGAACGCGTAGC
- Phenotype
-
MIM: 606628
MIM: 601498
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CNPY3-GNMT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141238034] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CNPY3-GNMT
- Gene Name
- CNPY3-GNMT readthrough
There are no transcripts associated with this gene.
- Gene
- GNMT
- Gene Name
- glycine N-methyltransferase
There are no transcripts associated with this gene.
- Gene
- PEX6
- Gene Name
- peroxisomal biogenesis factor 6
View Full Product Details