Product Details

SNP ID
rs148401284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41784609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGGGGAGGCAAGACAGGAGTAT[A/G]TGAAATAATTTGAGGAAATGCAAGA
Phenotype
MIM: 607744 MIM: 611389 MIM: 616168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FRS3 PubMed Links

Gene Details

Gene
FRS3
Gene Name
fibroblast growth factor receptor substrate 3
There are no transcripts associated with this gene.

Gene
PRICKLE4
Gene Name
prickle planar cell polarity protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013397.5 Intron NP_037529.3
Gene
TOMM6
Gene Name
translocase of outer mitochondrial membrane 6
There are no transcripts associated with this gene.

Gene
USP49
Gene Name
ubiquitin specific peptidase 49
There are no transcripts associated with this gene.

View Full Product Details