Product Details

SNP ID
rs148599364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:115942495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTTTCCAACGCAGTGTCTCAAA[G/T]GTAGGTCGTTCCTTAGGCTCTGCAT
Phenotype
MIM: 606573
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FRK PubMed Links

Gene Details

Gene
FRK
Gene Name
fyn related Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002031.2 2074 Silent Mutation ACA,ACC T479T NP_002022.1
XM_005266880.4 2074 Silent Mutation ACA,ACC T479T XP_005266937.1
XM_005266881.2 2074 Silent Mutation ACA,ACC T479T XP_005266938.1
XM_005266882.4 2074 Silent Mutation ACA,ACC T479T XP_005266939.1
XM_011535653.2 2074 Silent Mutation ACA,ACC T479T XP_011533955.1
XM_011535654.2 2074 Silent Mutation ACA,ACC T479T XP_011533956.1
XM_011535655.2 2074 Silent Mutation ACA,ACC T479T XP_011533957.1
XM_011535656.2 2074 Silent Mutation ACA,ACC T366T XP_011533958.1
XM_017010645.1 2074 Silent Mutation ACA,ACC T479T XP_016866134.1

View Full Product Details