Product Details

SNP ID
rs148620646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30346455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTGCAGGGGACAGCGCTGGACC[A/G]TACAGACCTGCCTAACATGCCAGCG
Phenotype
MIM: 612524 MIM: 605700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPP21 PubMed Links

Gene Details

Gene
RPP21
Gene Name
ribonuclease P/MRP subunit p21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199120.1 1319 Missense Mutation ATA,GTA I97V NP_001186049.1
NM_001199121.1 1319 Missense Mutation ATA,GTA I89V NP_001186050.1
NM_024839.2 1319 Missense Mutation ATA,GTA I89V NP_079115.1
Gene
TRIM39
Gene Name
tripartite motif containing 39
There are no transcripts associated with this gene.

Gene
TRIM39-RPP21
Gene Name
TRIM39-RPP21 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199119.1 1319 Missense Mutation ATA,GTA I438V NP_001186048.1

View Full Product Details