Product Details

SNP ID
rs148629293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29044408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTACCTGGTTGCAGGTACATGTA[C/G]ATAATAGTTCCATAGAACATAGACA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100129636 PubMed Links

Gene Details

Gene
LOC100129636
Gene Name
uncharacterized LOC100129636
There are no transcripts associated with this gene.

Gene
OR2W1
Gene Name
olfactory receptor family 2 subfamily W member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030903.3 768 Missense Mutation ATC,ATG I256M NP_112165.1

View Full Product Details