Product Details

SNP ID
rs149065283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31779018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGCGGCACCTTGACAGGATAGCC[C/T]GAGGCAGCACGGCGTTCCCGCAGAC
Phenotype
MIM: 192150 MIM: 609693
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VARS PubMed Links

Gene Details

Gene
VARS
Gene Name
valyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006295.2 4094 Silent Mutation TCA,TCG S1225S NP_006286.1
XM_005249362.2 4094 Silent Mutation TCA,TCG S1226S XP_005249419.1
XM_017011246.1 4094 Silent Mutation TCA,TCG S643S XP_016866735.1
XM_017011247.1 4094 Silent Mutation TCA,TCG S643S XP_016866736.1
Gene
VWA7
Gene Name
von Willebrand factor A domain containing 7
There are no transcripts associated with this gene.

View Full Product Details