Product Details

SNP ID
rs149139835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31580809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACACCCTCTCGCCCCTCCGGAGC[C/T]GCACCAGGCCGCCGAACCCCACGCT
Phenotype
MIM: 109170 MIM: 153440 MIM: 600978 MIM: 611550 MIM: 191160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100287329 PubMed Links

Gene Details

Gene
LOC100287329
Gene Name
uncharacterized LOC100287329
There are no transcripts associated with this gene.

Gene
LST1
Gene Name
leukocyte specific transcript 1
There are no transcripts associated with this gene.

Gene
LTA
Gene Name
lymphotoxin alpha
There are no transcripts associated with this gene.

Gene
LTB
Gene Name
lymphotoxin beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002341.1 597 Missense Mutation CAG,CGG Q212R NP_002332.1
NM_009588.1 597 UTR 3 NP_033666.1
Gene
NCR3
Gene Name
natural cytotoxicity triggering receptor 3
There are no transcripts associated with this gene.

Gene
TNF
Gene Name
tumor necrosis factor
There are no transcripts associated with this gene.

View Full Product Details