Product Details

SNP ID
rs149373880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:104943604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAAATTATTGATTATGAAAGTGG[C/T]GTTACAAATTTTTGGACAACTAAAG
Phenotype
MIM: 611044
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LIN28B PubMed Links

Gene Details

Gene
LIN28B
Gene Name
lin-28 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004317.3 Intron NP_001004317.1
XM_006715477.2 Intron XP_006715540.2
XM_011535818.2 Intron XP_011534120.1
Gene
LIN28B-AS1
Gene Name
LIN28B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details