Product Details

SNP ID
rs149409097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37444037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTAGTGGGTCTGATAGTGAGACC[A/G]AGGGGAAACAACACAGCTCTGACTC
Phenotype
MIM: 616189
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CMTR1 PubMed Links

Gene Details

Gene
CMTR1
Gene Name
cap methyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015050.2 425 Missense Mutation AAG,GAG K58E NP_055865.1
XM_005248955.3 425 Missense Mutation AAG,GAG K58E XP_005249012.1

View Full Product Details