Product Details
- SNP ID
-
rs149415241
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:53006333 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACTCACCAGGGGAAGGGTCTGGAA[C/T]AGGTGCTAGGTGTACCCTCTGCATA
- Phenotype
-
MIM: 612325
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ICK
PubMed Links
Gene Details
- Gene
- ICK
- Gene Name
- intestinal cell (MAK-like) kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014920.3 |
2906 |
Missense Mutation |
ATT,GTT |
I576V |
NP_055735.1 |
NM_016513.4 |
2906 |
Missense Mutation |
ATT,GTT |
I576V |
NP_057597.2 |
XM_011514419.1 |
2906 |
Missense Mutation |
ATT,GTT |
I583V |
XP_011512721.1 |
XM_011514420.2 |
2906 |
Missense Mutation |
ATT,GTT |
I583V |
XP_011512722.1 |
XM_011514421.1 |
2906 |
Missense Mutation |
ATT,GTT |
I583V |
XP_011512723.1 |
XM_017010485.1 |
2906 |
Missense Mutation |
ATT,GTT |
I583V |
XP_016865974.1 |
XM_017010486.1 |
2906 |
Missense Mutation |
ATT,GTT |
I583V |
XP_016865975.1 |
XM_017010487.1 |
2906 |
Missense Mutation |
ATT,GTT |
I583V |
XP_016865976.1 |
XM_017010488.1 |
2906 |
Missense Mutation |
ATT,GTT |
I576V |
XP_016865977.1 |
XM_017010489.1 |
2906 |
Missense Mutation |
ATT,GTT |
I576V |
XP_016865978.1 |
XM_017010490.1 |
2906 |
Missense Mutation |
ATT,GTT |
I576V |
XP_016865979.1 |
XM_017010491.1 |
2906 |
Missense Mutation |
ATT,GTT |
I576V |
XP_016865980.1 |
XM_017010492.1 |
2906 |
Missense Mutation |
ATT,GTT |
I576V |
XP_016865981.1 |
View Full Product Details