Product Details
- SNP ID
-
rs149481602
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:31587308 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGATCCCTGACCTAATGTTATC[A/G]CGGAATGATGGTAAGTAAAGTGTCT
- Phenotype
-
MIM: 109170
MIM: 600978
MIM: 611550
MIM: 191160
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LST1
PubMed Links
Gene Details
- Gene
- LST1
- Gene Name
- leukocyte specific transcript 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001166538.1 |
218 |
Silent Mutation |
TCA,TCG |
S3S |
NP_001160010.1 |
NM_007161.3 |
218 |
Silent Mutation |
TCA,TCG |
S3S |
NP_009092.3 |
NM_205837.2 |
218 |
Silent Mutation |
TCA,TCG |
S3S |
NP_995309.2 |
NM_205838.2 |
218 |
Silent Mutation |
TCA,TCG |
S3S |
NP_995310.2 |
NM_205839.2 |
218 |
Silent Mutation |
TCA,TCG |
S3S |
NP_995311.2 |
NM_205840.2 |
218 |
Silent Mutation |
TCA,TCG |
S3S |
NP_995312.2 |
XM_006715206.3 |
218 |
Silent Mutation |
TCA,TCG |
S3S |
XP_006715269.1 |
XM_006715209.3 |
218 |
Silent Mutation |
TCA,TCG |
S3S |
XP_006715272.1 |
XM_006715210.3 |
218 |
Silent Mutation |
TCA,TCG |
S3S |
XP_006715273.1 |
XM_011514914.2 |
218 |
Silent Mutation |
TCA,TCG |
S3S |
XP_011513216.1 |
- Gene
- LTB
- Gene Name
- lymphotoxin beta
There are no transcripts associated with this gene.
- Gene
- NCR3
- Gene Name
- natural cytotoxicity triggering receptor 3
There are no transcripts associated with this gene.
- Gene
- TNF
- Gene Name
- tumor necrosis factor
There are no transcripts associated with this gene.
View Full Product Details