Product Details

SNP ID
rs149521443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43620577 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATATATATATATATGCACAGAG[A/G]AACCTTAGCCAACAAATCCCTGAGT
Phenotype
MIM: 607434 MIM: 603968
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTPBP2 PubMed Links

Gene Details

Gene
GTPBP2
Gene Name
GTP binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286216.1 2930 UTR 3 NP_001273145.1
NM_019096.4 2930 UTR 3 NP_061969.3
XM_017010976.1 2930 Intron XP_016866465.1
Gene
MAD2L1BP
Gene Name
MAD2L1 binding protein
There are no transcripts associated with this gene.

Gene
POLH
Gene Name
DNA polymerase eta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291969.1 2930 Intron NP_001278898.1
NM_001291970.1 2930 Intron NP_001278899.1
NM_006502.2 2930 Intron NP_006493.1
XM_005249186.3 2930 Intron XP_005249243.1
XM_011514698.2 2930 Intron XP_011513000.1

View Full Product Details